Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • WNT11 Antibodies

          Invitrogen

          WNT11 Polyclonal Antibody

          3 Published Figures
          5 References
          View all (9) WNT11 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite WNT11 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (10)
          • Published Figures (3)
          WNT11 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          WNT11 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 13

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          WNT11 Antibody (PA5-21712) in ICC/IF

          WNT11 Polyclonal Antibody detects WNT11 protein at cytoplasm by immunofluorescent analysis. Sample: HeLa cells were fixed in ice-cold MeOH for 5 min. Green: WNT11 protein stained by WNT11 Polyclonal Antibody (Product # PA5-21712) diluted at 1:500. Blue: Hoechst 33342 staining. Scale bar = 10 µm. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          WNT11 Antibody in Immunocytochemistry (ICC/IF)
          WNT11 Antibody in Immunocytochemistry (ICC/IF)
          WNT11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          WNT11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          WNT11 Antibody in Western Blot (WB)
          WNT11 Antibody in Western Blot (WB)
          WNT11 Antibody in Western Blot (WB)
          WNT11 Antibody in Western Blot (WB)
          WNT11 Antibody in Western Blot (WB)
          WNT11 Antibody in Western Blot (WB)
          WNT11 Antibody in Immunohistochemistry (IHC)
          WNT11 Antibody in Western Blot (WB)
          WNT11 Antibody in Western Blot (WB)

          Product Details

          PA5-21712

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          View 3 publications 3 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:1,000
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:100-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein encompassing a sequence within the center region of human WNT11. The exact sequence is proprietary.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.07 mg/mL

          Amount

          107 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7, with 20% glycerol

          Contains

          0.025% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11154198

          Product Specific Information

          Recommended positive controls: 293T, A431, HeLa, HepG2, mouse heart.

          Predicted reactivity: Mouse (96%), Rat (96%), Xenopus laevis (81%), Rabbit (97%), Chicken (85%), Bovine (98%).

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          Ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Protein Wnt-11; unnamed protein product; wingless-related MMTV integration site 11; wingless-type MMTV integration site family, member 11; WNT11

          View more View less

          Gene Aliases: HWNT11; Wnt-11; WNT11

          View more View less

          UniProt ID: (Human) O96014, (Mouse) P48615

          View more View less

          Entrez Gene ID: (Human) 7481, (Rat) 140584, (Mouse) 22411

          View more View less

          Function(s)
          receptor binding frizzled binding cytokine activity protein binding protein kinase activator activity molecular_function transcription regulatory region DNA binding GTPase activator activity intercellular signal molecule
          Process(es)
          osteoblast differentiation kidney development epithelial to mesenchymal transition primary heart field specification secondary heart field specification outflow tract morphogenesis ventricular septum development atrial septum development apoptotic process signal transduction Wnt signaling pathway, calcium modulating pathway mesenchymal cell proliferation Wnt signaling pathway cell differentiation neuron differentiation bone mineralization adrenal gland development response to nutrient levels positive regulation of transforming growth factor beta2 production intracellular signal transduction non-canonical Wnt signaling pathway positive regulation of apoptotic process cell fate commitment maintenance of epithelial cell apical/basal polarity positive regulation of transcription, DNA-templated paraxial mesoderm formation notochord morphogenesis embryonic skeletal system development artery morphogenesis palate development convergent extension involved in axis elongation canonical Wnt signaling pathway Wnt signaling pathway, planar cell polarity pathway cloacal septation ventricular septum morphogenesis lung-associated mesenchyme development ureteric bud morphogenesis negative regulation of cartilage development somite development neuroendocrine cell differentiation secondary palate development bicellular tight junction assembly cellular response to mechanical stimulus cellular response to retinoic acid stem cell proliferation mesonephric duct development negative regulation of mesenchymal cell proliferation negative regulation of canonical Wnt signaling pathway negative regulation of fibroblast growth factor production epithelial cell apoptotic process positive regulation of epithelial cell apoptotic process negative regulation of stem cell proliferation protein phosphorylation positive regulation of gene expression negative regulation of cell growth positive regulation of cell migration negative regulation of cell migration protein localization to cell surface negative regulation of apoptotic process positive regulation of protein kinase activity negative regulation of transcription, DNA-templated positive regulation of stress fiber assembly negative regulation of cell death positive regulation of protein kinase C signaling cell-cell signaling multicellular organism development organ morphogenesis positive regulation of GTPase activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline