Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji.
Predicted reactivity: Mouse (92%), Rat (91%), Bovine (97%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PEF1 is a Ca(2+)-binding protein that belongs to the penta-EF-hand (PEF) protein family, which includes the calpain small subunit (CAPNS1), sorcin (SRI), grancalcin (GCA), and ALG2. PEF1 is a Ca(2+)-binding protein that belongs to the penta-EF-hand (PEF) protein family, which includes the calpain small subunit (CAPNS1), sorcin (SRI), grancalcin (GCA), and ALG2 (PDCD6) (Kitaura et al., 2001 ).
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory sperm binding protein; PEF; PEF protein with a long N-terminal hydrophobic domain; Peflin; Penta-EF hand domain-containing protein 1; unnamed protein product
Gene Aliases: ABP32; PEF1; PEF1A; UNQ1845/PRO3573
UniProt ID: (Human) Q9UBV8
Entrez Gene ID: (Human) 553115
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support