Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: 293T, A549, H1299, HCT116, Mouse brain.
Predicted reactivity: Mouse (95%), Rat (95%), Xenopus laevis (82%), Rabbit (94%), Bovine (95%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is likely a phosphoserine aminotransferase, based on similarity to proteins in mouse, rabbit, and Drosophila. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endometrial progesterone-induced protein; EPIP; Phosphohydroxythreonine aminotransferase; Phosphoserine aminotransferase; phosphoserine transaminase; PSAT; unnamed protein product
Gene Aliases: D8Ertd814e; EPIP; NLS2; PSA; PSAT; PSAT1; PSATD
UniProt ID: (Human) Q9Y617, (Mouse) Q99K85
Entrez Gene ID: (Human) 29968, (Mouse) 107272
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support