Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: SK-N-SH, SK-N-AS, mouse brain, human Ataxin3-transfected 293T.
Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (85%), Pig (99%), Chicken (89%), Sheep (98%), Bovine (98%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Machado-Joseph disease is an autosomal dominant neurologic disorder, and is now known to be the same as previously described spinocerebellar ataxia-3. MJD protein (Ataxin-3) contains (CAG)n repeats in the coding region, and the expansion of these repeats from the normal 13-36 to 68-79 is the cause of Machado-Joseph disease. There is a negative correlation between the age of onset and CAG repeat numbers. This protein interacts with key regulators (CBP, p300 and PCAF) of transcription and represses transcription, and also acts as a histone-binding protein that regulates transcription. MJD is a deubiquitinating enzyme.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ataxin-3; ataxin3; josephin; Machado-Joseph disease (spinocerebellar ataxia 3, olivopontocerebellar ataxia 3, autosomal dominant, ataxin 3) homolog; Machado-Joseph disease protein 1; Machado-Joseph disease protein 1 homolog; olivopontocerebellar ataxia 3; spin; Spinocerebellar ataxia type 3 protein
Gene Aliases: 2210008M02Rik; AI463012; AI647473; AT3; ataxin-3; ATX3; ATXN3; JOS; MJD; MJD1; SCA3; zgc:56323
UniProt ID: (Human) P54252, (Mouse) Q9CVD2
Entrez Gene ID: (Human) 4287, (Mouse) 110616, (Zebrafish) 394079
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support