Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
The target sequence has 91% sequence homology with rat.
JMJD1A is a histone demethylase that specifically demethylates 'Lys-9' of histone H3, thereby playing a central role in histone code. It preferentially demethylates mono- and dimethylated H3 'Lys-9' residue, with a preference for dimethylated residue, while it has weak or no activity on trimethylated H3 'Lys-9'. Demethylation of Lys residue generates formaldehyde and succinate. JMJD1A is involved in hormone-dependent transcriptional activation, by participating in recruitment to androgen-receptor target genes, resulting in H3 'Lys-9' demethylation and transcriptional activation. It's also Involved in spermatogenesis by regulating expression of target genes such as PRM1 and TMP1 which are required for packaging and condensation of sperm chromatin. Involved in obesity resistance through regulation of metabolic genes such as PPARA and UCP1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [histone H3]-dimethyl-L-lysine(9) demethylase 3A; DKFZp686A24246; DKFZp686P07111; JmjC domain-containing histone demethylation protein 2A; jumonji C domain-containing histone demethylase 2A; jumonji domain containing 1A; Jumonji domain-containing protein 1A; lysine (K)-specific demethylase 3A; Lysine-specific demethylase 3A; probable zinc finger protein; testis specific gene A; Testis-specific gene A protein; testis-specific protein A; unnamed protein product; Zinc finger protein TSGA
Gene Aliases: 1700105C21Rik; C230043E16Rik; JHDM2A; JHMD2A; JMJD1; JMJD1A; KDM2A; KDM3A; KIAA0742; TGSA; TSGA
UniProt ID: (Human) Q9Y4C1, (Mouse) Q6PCM1, (Rat) Q63679
Entrez Gene ID: (Human) 55818, (Mouse) 104263, (Rat) 312440
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support