Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
This antibody is predicted to react with rat based on sequence homology.
This gene encodes a member of the N-acetylglucosaminyltransferase family. The encoded protein is a beta-6-N-acetylglucosamine-transferase that catalyzes the formation of core 2 and core 4 O-glycans on mucin-type glycoproteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase 3; beta1,6-N-acetylglucosaminyltransferase-M; beta1,6GlcNAc-transferase; C2/4GnT; C2GnT-M; C2GnT-mucin type; core 2 beta-1,6-N-acetylglucosaminyltransferase II; Core 2/core 4 beta-1,6-N-acetylglucosaminyltransferase; glycosyltransferase; GnT-M; hC2GnT-M; Mucus-type core 2 beta-1,6-N-acetylglucosaminyltransferase; unnamed protein product
Gene Aliases: 2010013H22Rik; 2210021I22Rik; 2210401J11Rik; C2/4GnT; C24GNT; C2GNT2; C2GNTM; GCNT3; GNTM
UniProt ID: (Human) O95395, (Mouse) Q5JCT0
Entrez Gene ID: (Human) 9245, (Mouse) 72077
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support