Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 13
Recommended positive controls: HepG2, mouse kidney, rat kidney.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PCDGF is a glycoprotein and growth factor involved in the regulation of mitosis, early embryonic development, cell division, cell migration, cell survival, and hypothalamic differentiation. PCDGF is also known as progranulin because it is the precursor for smaller proteins known as epithelins or granulins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acrogranin; epithelin; Epithelin precursor; glycoprotein 88; Glycoprotein of 88 Kda; GP88; Granulin precursor; granulin-epithelin; granulins; PC cell-derived growth factor; PCDGF; PEPI; PGRN; Proepithelin; Progranulin; unnamed protein product
Gene Aliases: CLN11; FTD2; GEP; GP88; GRN; PCDGF; PEPI; PGRN
UniProt ID: (Human) P28799
Entrez Gene ID: (Human) 2896
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support