Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Recommended positive controls: HPCAL1 tansfected 293T cell, mouse brain.
Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (90%), Xenopus laevis (96%), Pig (99%), Chicken (98%), Sheep (99%), Rhesus Monkey (99%), Chimpanzee (99%), Bovine (99%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Hippocalcin-like protein 1, may be involved in the calcium-dependent regulation of rhodopsin phosphorylation.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcium-binding protein BDR-1; hippocalcin-like 1; visinin like 3; neural visinin-like 3; neural visinin-like protein 3; Hippocalcin-like protein 1; HLP2; HPCAL1; neural visinin-like 3; Neural visinin-like protein 3; NVL-3; unnamed protein product; VILIP-3; visinin like 3; Visinin-like protein 3
Gene Aliases: BDR1; HLP2; HPCAL1; NVP-3; Nvp3; VILIP-3; VILIP3; Vnsl3; Vsnl3
UniProt ID: (Human) P37235, (Mouse) P62748
Entrez Gene ID: (Human) 3241, (Mouse) 53602
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support