Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 6
Recommended positive controls: 293T, Neuro2A, C8D30, NIH-3T3, human BMPR1B-transfected 293T, Rat2.
Predicted reactivity: Mouse (98%), Rat (98%), Dog (98%), Pig (98%), Chicken (96%), Sheep (98%), Rhesus Monkey (99%), Bovine (99%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BMPR1B (also known as ALK6, CDw293) is a member of the transmembrane serine/threonine kinase that is the member of the bone morphogenetic protein (BMP) receptor, which is closely related to the activin receptors, ACVR1 and ACVR2. BMPR1B is mainly involved in the endochondral bone formation and embryogenesis. BMPR1B expressed in normal and cancerous prostate tissues and used in the endocrine therapy that given to the prostate cancer patients. BMPR1B receptor trafficking also play a significant role in FOP pathogenesis and used in human T-cell differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activin receptor-like kinase 6; ALK-6; BMP type-1B receptor; BMPR-1B; bone morphogenetic protein binding serine/threonine kinase receptor; bone morphogenetic protein receptor IB; Bone morphogenetic protein receptor type-1B; bone morphogenetic protein receptor, type IB; CDw293; serine/threonine receptor kinase; Serine/threonine-protein kinase receptor R6; SKR6; unnamed protein product
Gene Aliases: Acvrlk6; AI385617; ALK-6; ALK6; AMD3; AMDD; AV355320; BDA1D; BDA2; BMP15; BMPR-1B; BMPR-IB; BMPR1B; BMPRIB; CDw293; CFK-43a; SKR6
UniProt ID: (Human) O00238, (Mouse) P36898
Entrez Gene ID: (Human) 658, (Bovine) 407128, (Mouse) 12167, (Rat) 310914
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support