Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Recommended positive controls: Carboxypeptidase H tansfected 293T cell, mouse brain.
Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (85%), Xenopus laevis (83%), Pig (95%), Rhesus Monkey (99%), Chimpanzee (99%), Bovine (94%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a carboxypeptidase that cleaves C-terminal amino acid residues and is involved in the biosynthesis of peptide hormones and neurotransmitters, including insulin. It is a peripheral membrane protein. The protein specifically binds regulated secretory pathway proteins, including prohormones, but not constitutively secreted proteins. Mutations in this gene are implicated in type II diabetes.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: carbocypeptidase E; Carboxypeptidase E; Carboxypeptidase H; CarboxypeptidaseE; CarboxypeptidaseH; CBPE; cobalt-stimulated chromaffin granule carboxypeptidase; CPE; CPH; Cph 1; Enkephalin convertase; epididymis secretory sperm binding protein; insulin granule-associated carboxypeptidase; Prohormone-processing carboxypeptidase; unnamed protein product
Gene Aliases: BDVS; CPE; CPH; Cph-1; Cph1; fat; IDDHH; R74677
UniProt ID: (Human) P16870, (Mouse) Q00493
Entrez Gene ID: (Human) 1363, (Mouse) 12876
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support