Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: U87-MG.
Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (82%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DCUN1D4 contains 1 DCUN1 domain. The exact function of DCUN1D4 remains unknown.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DCN1, defective in cullin neddylation 1, domain containing 4; DCN1-like protein 4; DCNL4; DCUN1 domain-containing protein 4; Defective in cullin neddylation protein 1-like protein 4; KIAA0276; Squamous cell carcinoma-related oncogene 4; unnamed protein product
Gene Aliases: AI836376; DCNL4; DCUN1D4; KIAA0276; SCCRO4
UniProt ID: (Human) Q92564, (Mouse) Q8CCA0
Entrez Gene ID: (Human) 23142, (Mouse) 100737
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support