Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 6
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse liver.
Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (98%), Xenopus laevis (99%), Chicken (100%), Bovine (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene belongs to the SMC3 subfamily of SMC proteins. The encoded protein occurs in certain cell types as either an intracellular, nuclear protein or a secreted protein. The nuclear form, known as structural maintenance of chromosomes 3, is a component of the multimeric cohesin complex that holds together sister chromatids during mitosis, enabling proper chromosome segregation. Post-translational modification of the encoded protein by the addition of chondroitin sulfate chains gives rise to the secreted proteoglycan bamacan, an abundant basement membrane protein.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bamacan; Basement membrane-associated chondroitin proteoglycan; CG9802-PA; Chondroitin sulfate proteoglycan 6; chondroitin sulfate proteoglycan 6 (bamacan); Chromosome segregation protein SmcD; Chromosome-associated polypeptide; Chromosome-associated protein; cohesin subunit SMC3; hCAP; Mad member-interacting protein 1; SMC family; SMC protein 3; SMC3-PA; structural maintenace of chromosomes 3; Structural maintenance of chromosomes protein 3; unnamed protein product
Gene Aliases: BAM; Bamacan; BMH; CAP; CDLS3; CG9802; CSPG6; dCAP; Dmel\CG9802; Dmel_CG9802; DmSMC3; DmSMC3/Cap; dSMC3; HCAP; im:7142991; Mmip1; SMC-3; SMC3; SMC3L1; SmcD; wu:fb22e01; wu:fc30d07
UniProt ID: (Human) Q9UQE7, (Mouse) Q9CW03
Entrez Gene ID: (Fruit fly) 32627, (Human) 9126, (Mouse) 13006, (Zebrafish) 324475
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support