Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • SUV39H1 Antibodies

          Invitrogen

          SUV39H1 Polyclonal Antibody

          3 Published Figures
          2 References
          View all (10) SUV39H1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SUV39H1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          • Published Figures (3)
          SUV39H1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SUV39H1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SUV39H1 Antibody (PA5-29470) in ICC/IF

          Immunofluorescent analysis of SUV39H1 (green) in human hepatocytes. The cells were fixed, permeabilized and blocked. Cells were stained with a SUV39H1 polyclonal antibody (Product # PA5-29470) at a dilution of 1:500, followed by staining with Alexa fluor 488 goat anti-rabbit secondary antibody (Product # A-11008) at a dilution of 1:500 (green). Nuclei (blue) were counterstained with DAPI. Images were taken at 50X magnification. Data courtesy of the Antibody Data Exchange Program. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SUV39H1 Antibody in Immunocytochemistry (ICC/IF)
          SUV39H1 Antibody in Immunocytochemistry (ICC/IF)
          SUV39H1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SUV39H1 Antibody in Western Blot (WB)
          SUV39H1 Antibody in Immunocytochemistry (ICC/IF)
          SUV39H1 Antibody in Western Blot (WB)
          SUV39H1 Antibody in Western Blot (WB)

          Product Details

          PA5-29470

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:20,000
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:1,000
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein encompassing a sequence within the center region of human SUV39H1. The exact sequence is proprietary.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.52 mg/mL

          Amount

          52 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7, with 20% glycerol

          Contains

          0.025% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2546946

          Product Specific Information

          Recommended positive controls: 293T, A431, HeLa, HepG2, H1299, Raji, Mouse brain.

          Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (90%), Rhesus Monkey (100%), Bovine (100%).

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          This gene encodes an evolutionarily-conserved protein containing an N-terminal chromodomain and a C-terminal SET domain. The encoded protein is a histone methyltransferase that trimethylates lysine 9 of histone H3, which results in transcriptional gene silencing. Loss of function of this gene disrupts heterochromatin formation and may cause chromosome instability. Alternative splicing results in multiple transcript variants.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: H3-K9-HMTase 1; Histone H3-K9 methyltransferase 1; Histone-lysine N-methyltransferase SUV39H1; histone-lysine N-methyltransferase, H3 lysine-9 specific 1; located at OATL1; Lysine N-methyltransferase 1A; Position-effect variegation 3-9 homolog; pSUV39H; Su(var)3-9 homolog 1; Suppressor of variegation 3-9 homolog 1; Suv39h histone methyltransferase; SUV39H1; unnamed protein product

          View more View less

          Gene Aliases: AI852103; AL022883; DXHXS7466e; H3-K9-HMTase 1; KMT1A; MG44; mIS6; SUV39H; SUV39H1

          View more View less

          UniProt ID: (Human) O43463, (Mouse) O54864

          View more View less

          Entrez Gene ID: (Human) 6839, (Mouse) 20937

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II regulatory region sequence-specific DNA binding chromatin binding protein binding methyltransferase activity zinc ion binding S-adenosylmethionine-dependent methyltransferase activity transferase activity histone methyltransferase activity metal ion binding histone methyltransferase activity (H3-K9 specific) histone H3 methyltransferase activity histone H3K9me2 methyltransferase activity histone H3K9 trimethyltransferase activity protein methyltransferase activity histone-lysine N-methyltransferase activity protein N-terminus binding chromatin/chromatin-binding, or -regulatory protein histone modifying enzyme
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter chromatin silencing at rDNA blastocyst hatching regulation of DNA repair chromatin organization rRNA processing cellular response to DNA damage stimulus circadian rhythm determination of adult lifespan cell differentiation regulation of bone mineralization heterochromatin assembly methylation regulation of multicellular organism growth cellular response to glucose starvation epigenetic programming in the zygotic pronuclei negative regulation of cell cycle negative regulation of gene expression, epigenetic negative regulation of transcription, DNA-templated regulation of transcription by glucose rhythmic process cellular response to hypoxia energy homeostasis regulation of cellular senescence DNA packaging chromatin silencing transcription, DNA-templated regulation of transcription, DNA-templated protein methylation cell cycle chromatin modification histone lysine methylation histone H3-K9 dimethylation histone H3-K9 trimethylation negative regulation of circadian rhythm histone H3-K9 methylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline