Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 6
Recommended positive controls: 293T, A431, mouse brain, rat brain.
Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (93%), Chicken (98%), Rhesus Monkey (100%), Bovine (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a syntaxin-binding protein. The encoded protein appears to play a role in release of neurotransmitters via regulation of syntaxin, a transmembrane attachment protein receptor. Mutations in this gene have been associated with infantile epileptic encephalopathy-4. Alternatively spliced transcript variants have been described.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EIEE4; FLJ37475; hUNC18; MGC146482; minisatellite 10g detected by probe MMS10; munc-18c; MUNC18-1; N-Sec1; neuronal SEC1; p67; Protein unc-18 homolog 1; Protein unc-18 homolog A; rbSec1; rop; RP11-56D16.3; Syntaxin-binding protein 1; UNC-18 homolog; unc-18-1; Unc-18A; Unc18-1; unnamed protein product
Gene Aliases: AI317162; AI326233; ANC18HA; DEE4; MMS10-G; Ms10g; Munc-18a; MUNC18-1; N-Sec1; NSEC1; NSEC1A; P67; Rb-sec1; RBSEC1; rbSec1A; rbSec1B; Sec1; STXBP1; Sxtbp1; unc-18A; UNC18; Unc18-1; UNC18A; Unc18h
UniProt ID: (Human) P61764, (Mouse) O08599, (Rat) P61765
Entrez Gene ID: (Human) 6812, (Mouse) 20910, (Rat) 25558
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support