Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: 293T, A431, Jurkat, Raji.
Predicted reactivity: Mouse (98%), Rat (98%), Chicken (91%), Bovine (95%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Members of the makorin family, including MKRN2, have a characteristic zinc finger composition that suggests that they are ribonucleoproteins (Gray et al., 2001).
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase makorin-2; makorin RING zinc-finger protein 2; probable E3 ubiquitin-protein ligase makorin-2; RING finger protein 62; RING zinc-finger protein; RING-type E3 ubiquitin transferase makorin-2; unnamed protein product
Gene Aliases: HSPC070; MKRN2; RNF62
UniProt ID: (Human) Q9H000
Entrez Gene ID: (Human) 23609
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support