Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: 293T.
Predicted reactivity: Mouse (89%), Rat (90%), Bovine (92%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RNase P, which consists of a complex of an RNA species (H1 RNA), POP1 (Processing of Precursor 1), POP5 (Processing of Precursor 5), and at least seven Rpps (including Rpp14, Rpp29, Rpp30 and Rpp38), removes the 5' leader sequences from precursor tRNA molecules. The structurally related RNase MRP plays an essential role in the formation of the 5' end of 5.8S rRNA. Both RNase P and RNase MRP are associated with Th/To ribonucleoproteins; Rpp30 and Rpp38 have specifically been implicated as Th autoantigens which contribute to the autoimmune disease systemic sclerosis.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ribonuclease P protein subunit p30; ribonuclease P/MRP 30kDa subunit; RNase P subunit 2; RNaseP protein p30; unnamed protein product
Gene Aliases: RNASEP2; RPP30; TSG15
UniProt ID: (Human) P78346
Entrez Gene ID: (Human) 10556
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support