Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: 293T.
Predicted reactivity: Mouse (91%), Rat (93%), Xenopus laevis (82%), Pig (97%), Rhesus Monkey (99%), Bovine (96%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein subunit of the GINS heterotetrameric complex, which is essential for the initiation of DNA replication and replisome progression in eukaryotes. Alternatively spliced transcript variants encoding distinct isoforms have been described.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA replication complex GINS protein PSF3; FLJ13912; GINS complex subunit 3; GINS complex subunit 3 (Psf3 homolog); GINS complex subunit 3 (Psf3 homolog) isoform GINS3_1; GINS complex subunit 3 (Psf3 homolog) isoform GINS3_2; GINS complex subunit 3 (Psf3 homolog) isoform GINS3_3; GINS complex subunit 3 (Psf3 homolog) isoform GINS3_4; unnamed protein product
Gene Aliases: GINS3; PSF3
UniProt ID: (Human) Q9BRX5
Entrez Gene ID: (Human) 64785
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support