Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: mouse Liver.
Predicted reactivity: Mouse (81%), Rat (81%), Rhesus Monkey (95%), Bovine (82%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Antibodies against the protein encoded by this gene were found only in plasma from cancer patients. While it may be a target for immunotherapy, the function of this gene is unknown.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer/testis antigen 112; family with sequence similarity 46 member D; Non-canonical poly(A) polymerase FAM46D; protein FAM46D; putative nucleotidyltransferase FAM46D; Terminal nucleotidyltransferase 5D; unnamed protein product
Gene Aliases: CT1.26; CT112; FAM46D; Gm377; TENT5D
UniProt ID: (Human) Q8NEK8
Entrez Gene ID: (Mouse) 213449, (Human) 169966
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support