Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7, NIH-3T3.
Predicted reactivity: Mouse (82%), Rat (82%), Rhesus Monkey (95%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The TTLL10 gene encodes a protein that functions as a polyglycylase, playing an important role in the modification of nucleosome assembly protein 1 (NAP1) both in vivo and in vitro. This enzymatic activity is dependent on the presence of ATP and is abolished in mutant forms where the glutamate in the active site is replaced by valine. TTLL10 specifically incorporates glycine into recombinant NAP1 at Glu residues 359 and 360. This modification process likely has implications for protein interactions and cellular processes involving NAP1. Additionally, TTLL10 can perform tubulin glycylation when co-expressed with TTLL8, suggesting a complex interplay between these enzymes in cellular microtubule dynamics.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Inactive polyglycylase TTLL10; Protein polyglycylase TTLL10; tubulin tyrosine ligase-like family, member 10; tubulin tyrosine ligase-like family, member 5; Tubulin--tyrosine ligase-like protein 10; unnamed protein product
Gene Aliases: 4833412E22Rik; 4930595O22Rik; AU022799; BC042775; TTLL10; TTLL5
UniProt ID: (Human) Q6ZVT0, (Mouse) A4Q9F3
Entrez Gene ID: (Human) 254173, (Mouse) 330010
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support