Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset (100%) Monkey, Mouse, Rat, Hamster (95%) Bovine, Rabbit, Horse (90%).
Mediates thiol-dependent retention in the early secretory pathway, forming mixed disulfides with substrate proteins through its conserved CRFS motif. Inhibits the calcium channel activity of ITPR1. May have a role in the control of oxidative protein folding in the endoplasmic reticulum. Required to retain ERO1L and ERO1LB in the endoplasmic reticulum.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endoplasmic reticulum resident protein 44; endoplasmic reticulum resident protein 44 kDa; epididymis secretory sperm binding protein; ER protein 44; OTTHUMP00000063799; protein disulfide isomerase family A, member 10; thioredoxin domain containing 4 (endoplasmic reticulum); Thioredoxin domain-containing protein 4; unnamed protein product
Gene Aliases: ERP44; KIAA0573; PDIA10; TXNDC4; UNQ532/PRO1075
UniProt ID: (Human) Q9BS26
Entrez Gene ID: (Human) 23071
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support