Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Marmoset (87%) Mouse, Rat, Dog, Elephant, Horse (80%).
QSOX2 is a member of the sulfhydryl oxidase/quiescin-6 (Q6) family (QSOX1) that regulates the sensitization of neuroblastoma cells for IFN-gamma (IFNG)-induced cell death.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neuroblastoma-derived sulfhydryl oxidase; quiescin Q6 sulfhydryl oxidase 2; quiescin Q6-like 1; Quiescin Q6-like protein 1; Sulfhydryl oxidase 2; thiol oxidase 2; unnamed protein product
Gene Aliases: QSCN6L1; QSOX2; SOXN
UniProt ID: (Human) Q6ZRP7
Entrez Gene ID: (Human) 169714
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support