Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • AGTR2 Antibodies

          Invitrogen

          AGTR2 Polyclonal Antibody

          View all (13) AGTR2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite AGTR2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          AGTR2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          AGTR2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          AGTR2 Antibody (PA5-33341) in IHC (P)

          Immunohistochemistry analysis of AGTR2 in Human Prostate. Samples were incubated with AGTR2 polyclonal antibody (Product # PA5-33341). formalin-fixed paraffin-embedded . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          AGTR2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AGTR2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AGTR2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AGTR2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AGTR2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AGTR2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))

          Product Details

          PA5-33341

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Paraffin) (IHC (P))

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Bat, Bovine, Dog, Horse, Hamster, Human, Mouse, Non-human primate, Sheep, Pig, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic 16 amino acid peptide from internal region of human AGTR2.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          0.1% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2550723

          Product Specific Information

          Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Sheep, Dog, Bat, Bovine, Hamster, Elephant, Panda, Horse, Pig (100%).

          For IHC(P), use proteinase K antigen retrieval. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

          Target Information

          Angiotensin II is a potent vasopressor hormone and a primary regulator of aldosterone secretion that acts through at least two types of receptors, AGTR1 and AGTR2. It is an important effector controlling blood pressure and volume in the cardiovascular system and plays a major role in the development of the mammalian kidney and urinary tract. Like AGTR1, AGTR2 is a seven transmembrane G protein-coupled receptor (GPCR), but AGTR2 does not demonstrate most of the classic features of GPCR signaling. AGTR2 is involved in a wide range of activities, including the induction of neurite outgrowth and the inhibition of cellular proliferation, in addition to the known function of mediation of vasoconstriction.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: AGTR2; Angiotensin ii at2; angiotensin II receptor, type 2; angiotensin II subtype 2 receptor, AT2; angiotensin II type 2 receptor; Angiotensin II type-2 receptor; angiotensin receptor 2; angiotensin type II receptor; AT2; AT2 receptor; Type-2 angiotensin II receptor; type-2 angiotensin II receptor-like protein; unnamed protein product

          View more View less

          Gene Aliases: AGTR2; AI316812; AT2; AT2-R; AT2R; AT2R; ATGR2; AW107640; MRX88

          View more View less

          UniProt ID: (Human) P50052, (Sheep) Q28929, (Rat) P35351, (Mouse) P35374

          View more View less

          Entrez Gene ID: (Human) 186, (Sheep) 443335, (Pig) 733586, (Dog) 403835, (Horse) 100053878, (Bovine) 407157, (Chinese hamster) 100750901, (Rat) 24182, (Mouse) 11609

          View more View less

          Function(s)
          G-protein coupled receptor activity angiotensin type II receptor activity protein binding receptor antagonist activity G-protein alpha-subunit binding transcription factor binding peptide hormone binding signal transducer activity G-protein coupled receptor
          Process(es)
          blood vessel remodeling regulation of systemic arterial blood pressure by circulatory renin-angiotensin vasodilation by angiotensin involved in regulation of systemic arterial blood pressure brain renin-angiotensin system inflammatory response signal transduction cell surface receptor signaling pathway G-protein coupled receptor signaling pathway G-protein coupled receptor signaling pathway coupled to cGMP nucleotide second messenger brain development regulation of blood pressure negative regulation of heart rate negative regulation of cell growth regulation of metanephros size exploration behavior nitric oxide-cGMP-mediated signaling pathway angiotensin-activated signaling pathway vasodilation regulation of apoptotic process negative regulation of blood vessel endothelial cell migration positive regulation of transcription, DNA-templated negative regulation of neurotrophin TRK receptor signaling pathway neuron apoptotic process positive regulation of metanephric glomerulus development positive regulation of branching involved in ureteric bud morphogenesis regulation of blood vessel diameter positive regulation of extrinsic apoptotic signaling pathway kidney development renin-angiotensin regulation of aldosterone production cellular sodium ion homeostasis positive regulation of cell proliferation response to organonitrogen compound negative regulation of norepinephrine secretion cerebellar cortex development neuron differentiation negative regulation of icosanoid secretion positive regulation of phosphoprotein phosphatase activity intracellular signal transduction positive regulation of renal sodium excretion aldosterone secretion dopamine biosynthetic process regulation of transcription factor import into nucleus negative regulation of neuron apoptotic process positive regulation of vasodilation negative regulation of fibroblast proliferation positive regulation of cytokine secretion positive regulation of nitric-oxide synthase activity kidney morphogenesis cell growth involved in cardiac muscle cell development cellular response to dexamethasone stimulus regulation of vasodilation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline