Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset, Dog, Panda, Rabbit, Pig (100%) Gibbon, Bovine, Elephant, Horse (95%).
GPRs are characterized by the presence of 7 transmembrane domains. GPR133 is an orphan receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adhesion G-protein coupled receptor D1; DKFZp434B1272; FLJ16770; G protein-coupled receptor 133; G-protein coupled receptor 133; G-protein coupled receptor PGR25; MGC138512; MGC138514; probable G-protein coupled receptor 133; unnamed protein product
Gene Aliases: ADGRD1; GPR133; PGR25
UniProt ID: (Human) Q6QNK2
Entrez Gene ID: (Pig) 100736874, (Dog) 486229, (Human) 283383, (Rabbit) 100352052
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support