Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The Latrophilin family of G protein-coupled receptors consists of three members, latrophilin-1, 2 and 3, each of which displays distinct tissue distribution and function. Latrophilin-1, the most characterized member of this family, acts as a receptor for a-latrotoxin, a component of venom from the black widow spider. Binding of a-latrotoxin to latrophilin-1 triggers synaptic vesicle exocytosis via both Ca2+-dependent and -independent mechanisms, resulting in vesicle depletion. Latrophilin-1 is abundant in brain and present in endocrine cells. Latrophilin-3 is also brain-specific, whereas latrophilin-2 expression is ubiquitous.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adhesion G protein-coupled receptor L2; Calcium-independent alpha-latrotoxin receptor 2; calcium-independent alpha-latrotoxin receptor homolog 2; CIRL-2; Cl2aa; G-protein coupled receptor; latrophilin 1; latrophilin 2; Latrophilin homolog 1; Latrophilin-2; lectomedin 2; Lectomedin-1; unnamed protein product
Gene Aliases: ADGRL2; AI450192; Cirl-2; CIRL2; CL2; Cl2ac; Gm619; KIAA0786; LEC1; LEC2; LPH2; LPHH1; LPHN2; mKIAA0786
UniProt ID: (Human) O95490, (Bovine) O97817, (Rat) O88923
Entrez Gene ID: (Horse) 100052932, (Rabbit) 100343149, (Human) 23266, (Dog) 490193, (Bovine) 281278, (Pig) 100524685, (Mouse) 99633, (Rat) 171447
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support