Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
A suggested positive control is A-20 cell lysate.
PA5-34544 can be used with blocking peptide PEP-1587.
The solute carrier family 27 (fatty acid transporter), member 6 (SLC27A6) is a member of the fatty acid transport protein family (FATP), which are involved in the uptake of long-chain fatty acids. SLC27A6 is principally expressed in the heart on the sarcolemma and colocalizes with CD36, another molecule involved in the uptake of long chain fatty acids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acyl-CoA synthetase very long chain family, member 2; Arachidonate--CoA ligase; FATP-6; Fatty-acid-coenzyme A ligase, very long-chain 2; Long-chain fatty acid transport protein 6; Long-chain-fatty-acid--CoA ligase; SLC27A6; solute carrier family 27 (fatty acid transporter), member 6; Solute carrier family 27 member 6; unnamed protein product; Very long-chain acyl-CoA synthetase homolog 1; VLACS2; VLCSH1
Gene Aliases: 4732438L20Rik; ACSVL2; FACVL2; Fatp1; FATP6; SLC27A6; VLCS-H1
UniProt ID: (Human) Q9Y2P4, (Mouse) E9Q9W4
Entrez Gene ID: (Human) 28965, (Rat) 291582, (Mouse) 225579
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support