Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: 293T, A431, Molt-4, Raji.
Predicted reactivity: Human (99%), Rhesus Monkey (96%), Bovine (84%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GK5 (Glycerol Kinase 5) is a Protein Coding gene. Diseases associated with GK5 include Type 1 Diabetes Mellitus 7 and Type 1 Diabetes Mellitus 13. Among its related pathways are glycerol degradation and triacylglycerol biosynthesis. Gene Ontology (GO) annotations related to this gene include phosphotransferase activity, alcohol group as acceptor and glycerol kinase activity. An important paralog of this gene is GK. [GeneCards]
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP:glycerol 3-phosphotransferase 5; GK 5; glycerokinase 5; Glycerol kinase 5; glycerol kinase 5 (putative); unnamed protein product
Gene Aliases: GK5
UniProt ID: (Human) Q6ZS86
Entrez Gene ID: (Human) 256356
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support