Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
May be involved in maintaining Golgi structure.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 88 kDa Golgi matrix protein; 88-kDa golgi protein; GM88; GM88 autoantigen; golgi autoantigen, golgin subfamily a, 8A; golgi protein; Golgin subfamily A member 8A; Golgin subfamily A member 8B; golgin-67
Gene Aliases: CFAP286; FAP286; GM88; GOLGA8A; GOLGA8B; KIAA0855
UniProt ID: (Human) A7E2F4
Entrez Gene ID: (Human) 23015
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support