Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 10
This antibody is predicted to react with mouse based on sequence homology.
This intronless gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in tumor formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BF-2; Brain factor 2; Forkhead box protein D1; FORKHEAD BOX PROTEIN D1 (FORKHEAD-RELATED PROTEIN FKHL8) (FORKHEAD-RELATED TRANSCRIPTION FACTOR 4) (FREAC-4); Forkhead, drosophila, homolog-like 8; forkhead-like 8; forkhead-related activator 4; Forkhead-related protein FKHL8; Forkhead-related transcription factor 4; FREAC-4; HFH-BF-2; HNF-3/forkhead homolog, brain factor 2
Gene Aliases: AI385632; BF-2; FKHL8; FOXD1; FREAC-4; FREAC4; Hfh10; Hfhbf2
UniProt ID: (Human) Q16676, (Mouse) Q61345
Entrez Gene ID: (Human) 2297, (Mouse) 15229
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support