Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Aquaporin 2 Antibodies

          Invitrogen

          Phospho-Aquaporin 2 (Ser264) Polyclonal Antibody

          Advanced Verification
          2 Published Figures
          1 Reference
          View all (32) Aquaporin 2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Phospho-Aquaporin 2 (Ser264) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (2)
          • Advanced Verification (1)
          Phospho-Aquaporin 2 (Ser264) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-Aquaporin 2 (Ser264) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-Aquaporin 2 (Ser264) Antibody (PA5-35387) in WB

          Western blot of Aquaporin 2 in rat kidney lysate showing specific immunolabeling of a bands at ~29 kDa and 37 kDa corresponding to glycosylated form of the Phospho-Aquaporin 2 (Ser264) polyclonal antibody (Product # PA5-35387) in the vasopressin (AVP) treated lane (+), but not in the control lane (-). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-Aquaporin 2 (Ser264) Antibody in Western Blot (WB)
          Phospho-Aquaporin 2 (Ser264) Antibody in Western Blot (WB)
          Phospho-Aquaporin 2 (Ser264) Antibody in Western Blot (WB)
          Phospho-Aquaporin 2 (Ser264) Antibody
          Phospho-Aquaporin 2 (Ser264) Polyclonal Antibody

          Product Details

          PA5-35387

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser264 of rat AQP2 conjugated to KLH
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          0.01M HEPES, pH 7.5, with 0.15M NaCl, 50% glycerol, 0.1mg/mL BSA

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2552697

          Product Specific Information

          This antibody is predicted to react with canine, bovine, chicken, human, mouse and non-human primate based on 100% sequence homology.

          PA5-35387 is specific for the ~29kDa AQP2 protein phosphorylated at Ser264, and also recognizes the glycosylated form of AQP2 at ~37kDa. This antibody contains enough material to conduct 10 mini-Western blots.

          Target Information

          Aquaporin 2 (AQP2) is a hormonally regulated water channel located in the renal collecting duct. Mutations in the AQP2 gene cause hereditary nephrogenic diabetes insipidus in humans. A vasopressin induced cAMP increase results in the phosphorylation of AQP2 at serine-256 and its translocation from the intracellular vesicles to the apical membrane of principal cells. Recently, serine-261 has been identified as a novel phosphorylation site on AQP2 and levels of phosphorylated S261 have been shown to decrease with vasopressin treatment suggesting its involvement in vasopressin-dependent AQP2 trafficking.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ADH water channel; AQP 2; AQP CD; AQP-2; AQP-CD; AQP2; AQPCD; aquaporin 2 (collecting duct); Aquaporin CD; Aquaporin-2; Aquaporin-CD; Aquaporin2; Aquaporine 2; Collecting duct water channel protein; MGC34501; Water channel protein for renal collecting duct; water-channel aquaporin 2; WCH CD; WCH-CD; WCHCD

          View more View less

          Gene Aliases: AQP-2; AQP-CD; AQP2; aquaporin-2; cph; jpk; NDI2; WCH-CD

          View more View less

          UniProt ID: (Human) P41181, (Mouse) P56402, (Rat) P34080

          View more View less

          Entrez Gene ID: (Human) 359, (Mouse) 11827, (Rat) 25386

          View more View less

          Function(s)
          water transmembrane transporter activity protein binding glycerol transmembrane transporter activity water channel activity channel activity actin binding transporter activity glycerol channel activity PDZ domain binding transporter
          Process(es)
          renal water homeostasis renal water transport water transport actin filament organization glycerol transport cellular response to water deprivation protein homotetramerization transmembrane transport cellular response to copper ion cellular response to mercury ion metanephric collecting duct development transport cell volume homeostasis apoptotic process cellular water homeostasis actin filament depolymerization carbohydrate transmembrane transport ion transmembrane transport positive regulation of calcium ion transport hyperosmotic response female pregnancy aging response to water deprivation response to salt stress response to hormone response to lithium ion water homeostasis response to lipopolysaccharide response to glucagon response to starvation response to calcium ion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline