Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Predicted reactivity: Mouse (94%), Rat (93%), Pig (92%), Chicken (82%), Rhesus Monkey (100%), Bovine (91%).
The METTL11A gene encodes an N-terminal methyltransferase for the RAN (MIM 601179) guanine nucleotide exchange factor regulator of chromosome condensation 1 (RCC1; MIM 179710). METTL11A enzyme alpha-N-methylates other protein targets such as SET (MIM 600960) and RB (MIM 180200).
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha N-terminal protein methyltransferase 1A; Methyltransferase-like protein 11A; N-terminal RCC1 methyltransferase; N-terminal Xaa-Pro-Lys N-methyltransferase 1; NTM1A; unnamed protein product; X-Pro-Lys N-terminal protein methyltransferase 1A
Gene Aliases: AD-003; C9orf32; HOMT1A; METTL11A; NRMT; NRMT1; NTM1A; NTMT1
UniProt ID: (Human) Q9BV86
Entrez Gene ID: (Human) 28989
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support