Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Recommended positive controls: MDA-MB-231, NT2D1, human Rubicon-transfected 293T.
Predicted reactivity: Mouse (92%), Pig (94%), Bovine (93%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Two Beclin-1-interacting proteins, the run domain Beclin-1 interacting and cysteine-rich containing protein (Rubicon) and ATG14L, reciprocally regulate autophagy at different stages. Knockdown of Rubicon caused enhancement of autophagy while that of ATG14L caused a defect in autophagosome formation. Rubicon functions as part of a Beclin-1-PIK3C3-containing autophagy complex and is also an essential, positive regulator of the NAPDH oxidase complex. Upon microbial infection or TLR2 activation, Rubicon interacts with the CYBA subunit of the NAPDH oxidase complex, leading to a burst of reactive oxygen species and inflammatory cytokines.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Baron; Beclin-1 associated RUN domain containing protein; Rubicon; RUN and cysteine rich domain containing beclin 1 interacting protein; RUN domain and cysteine-rich domain containing, Beclin 1-interacting protein; run domain Beclin-1 interacting and cystein-rich containing protein; run domain Beclin-1 interacting and cysteine-rich containing protein; Run domain Beclin-1-interacting and cysteine-rich domain-containing protein; rundataxin; unnamed protein product
Gene Aliases: 5330403K09; KIAA0226; mKIAA0226; RUBCN; RUBICON; SCAR15
UniProt ID: (Mouse) Q80U62, (Human) Q92622
Entrez Gene ID: (Mouse) 100502698, (Human) 9711
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support