Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FOXC1/FOXC2 Antibodies

          Invitrogen

          FOXC1/FOXC2 Polyclonal Antibody

          View all (2) FOXC1/FOXC2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite FOXC1/FOXC2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          FOXC1/FOXC2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FOXC1/FOXC2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FOXC1/FOXC2 Antibody (PA5-36261) in IHC (P)

          Immunohistochemistry analysis of FOXC1/FOXC2 in paraffin-embedded human tonsil carcinoma tissue. Samples were incubated with FOXC1/FOXC2 polyclonal antibody (Product # PA5-36261) at a dilution of 1:50. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FOXC1/FOXC2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FOXC1/FOXC2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FOXC1/FOXC2 Antibody in Western Blot (WB)
          FOXC1/FOXC2 Antibody in Western Blot (WB)
          FOXC1/FOXC2 Antibody in Western Blot (WB)
          FOXC1/FOXC2 Polyclonal Antibody

          Product Details

          PA5-36261

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to amino acids 100-150 of human FoxC1
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.2, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2553446

          Product Specific Information

          This antibody detects endogenous protein at a molecular weight of 57 kDa.

          Purity is >95% by SDS-PAGE.

          Target Information

          The forkhead transcription factor genes FOXC1 (Mf1) and FOXC2 (Mfh1) interact with the Notch signaling pathway and are required for the prepatterning of anterior and posterior domains in the presumptive somites through a putative Notch/Delta/Mesp regulatory loop. The genes have similar, dose-dependent functions, and compensate for each other in the early development of the heart, blood vessels and somites. Both FOXC1 and FOXC2 are expressed in the mesenchyme from which the ocular drainage structures derive. FOXC1 and FOXC2 also interact in kidney and heart development. Mutations in the FOXC1 gene result in Axenfeld-Rieger malformations of the anterior segment of the eye and lead to an increased susceptibility of glaucoma, including juvenile glaucoma. Functional regions in FOXC1 are required for nuclear localization and transcriptional regulation. Specifically, two regions in the FOXC1 forkhead domain, one rich in basic amino acid residues, and a second, highly conserved among all FOX proteins, are necessary for nuclear localization of the FOXC1 protein.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: BF-3; Brain factor 3; congenital hydrocephalus; forkhead box C1 protein; forkhead box C2 (MFH-1 mesenchyme forkhead 1); forkhead box C2 (MFH-1, mesenchyme forkhead 1); Forkhead box protein C1; Forkhead box protein C2; forkhead homolog 14; forkhead, Drosophila, homolog-like 14; forkhead, drosophila, homolog-like 7; forkhead-related activator 3; Forkhead-related protein FKHL14; Forkhead-related protein FKHL7; Forkhead-related transcription factor 3; forkhead/winged helix-like transcription factor 7; FOXC1; FREAC-3; HFH-BF-3; Mesenchyme fork head protein 1; mesenchyme forkhead 1; Mesoderm/mesenchyme forkhead 1; MF-1; MFH-1 protein; MFH-1,mesenchyme forkhead 1; myeloid factor-delta; Transcription factor FKH-1; Transcription factor FKH-14

          View more View less

          Gene Aliases: ARA; ASGD3; ch; fkh-1; Fkh1; Fkh14; FKHL14; FKHL7; FOXC1; FOXC2; FREAC-3; FREAC3; frkhda; Hfhbf3; IGDA; IHG1; IRID1; LD; Mf1; Mf4; MFH-1; MFH1; RIEG3

          View more View less

          UniProt ID: (Human) Q12948, (Human) Q99958, (Mouse) Q61572, (Rat) Q63246, (Mouse) Q61850

          View more View less

          Entrez Gene ID: (Human) 2296, (Human) 2303, (Mouse) 17300, (Rat) 364706, (Rat) 171356, (Mouse) 14234

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding DNA binding, bending sequence-specific DNA binding RNA polymerase II sequence-specific DNA binding transcription factor binding DNA-binding transcription factor binding promoter-specific chromatin binding bacterial-type RNA polymerase transcriptional activator activity, sequence-specific DNA binding chromatin DNA binding identical protein binding sequence-specific double-stranded DNA binding transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding transcription factor binding transcription regulatory region DNA binding molecular_function transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding winged helix/forkhead transcription factor
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter skeletal system development ossification angiogenesis ovarian follicle development blood vessel development eye development ureteric bud development in utero embryonic development somitogenesis kidney development lymph vessel development endochondral ossification blood vessel remodeling heart morphogenesis apoptotic process involved in outflow tract morphogenesis regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter Notch signaling pathway brain development heart development cell proliferation germ cell migration anatomical structure morphogenesis positive regulation of gene expression positive regulation of epithelial to mesenchymal transition mesenchymal cell development neural crest cell development cell migration negative regulation of angiogenesis cerebellum development cell differentiation collagen fibril organization glycosaminoglycan metabolic process lacrimal gland development embryonic heart tube development maintenance of lens transparency vascular endothelial growth factor signaling pathway odontogenesis of dentin-containing tooth camera-type eye development positive regulation of DNA binding positive regulation of keratinocyte differentiation positive regulation of transcription, DNA-templated negative regulation of mitotic cell cycle positive regulation of transcription from RNA polymerase II promoter regulation of organ growth vascular endothelial growth factor receptor signaling pathway paraxial mesoderm formation mesenchymal cell differentiation artery morphogenesis ventricular cardiac muscle tissue morphogenesis cardiac muscle cell proliferation chemokine-mediated signaling pathway cellular response to epidermal growth factor stimulus glomerular epithelium development regulation of blood vessel diameter negative regulation of lymphangiogenesis positive regulation of hematopoietic progenitor cell differentiation positive regulation of hematopoietic stem cell differentiation negative regulation of apoptotic process involved in outflow tract morphogenesis positive regulation of core promoter binding cellular response to chemokine patterning of blood vessels metanephros development lymphangiogenesis mesoderm development insulin receptor signaling pathway response to hormone positive regulation of endothelial cell migration positive regulation of cell adhesion mediated by integrin positive regulation of vascular wound healing paraxial mesodermal cell fate commitment embryonic cranial skeleton morphogenesis embryonic viscerocranium morphogenesis embryonic skeletal system morphogenesis glomerular endothelium development glomerular visceral epithelial cell differentiation glomerular mesangial cell development positive regulation of cell migration involved in sprouting angiogenesis negative regulation of cold-induced thermogenesis transcription, DNA-templated transcription from RNA polymerase II promoter regulation of blood vessel size biological_process multicellular organism development positive regulation of integrin activation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline