Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide sequence: CVCVRKRPLN QRETTLKDLD IITVPSDNVV MVHESKQKVD LTRYLQNQTF
Sequence homology: Dog: 87%; Guinea Pig: 87%; Horse: 79%; Human: 100%; Mouse: 87%; Rabbit: 80%; Rat: 80%
KIF2B is the plus end-directed microtubule-dependent motor required for spindle assembly and chromosome movement. KIF2B has microtubule depolymerization activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kinesin protein; Kinesin-like protein KIF2B; NYD-KIF2; testis tissue sperm-binding protein Li 82P; unnamed protein product
Gene Aliases: KIF2B
UniProt ID: (Human) Q8N4N8
Entrez Gene ID: (Human) 84643
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support