Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide sequence: VHMAEEIFTC MEGWKDLPAT SCLLQHQGPQ SEWKPYRDTE DREAFQTGQN
Sequence homology: Human: 100%
ZNF548 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 11 C2H2-type zinc fingers and 1 KRAB domain. ZNF548 may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: unnamed protein product; Zinc finger protein 548
Gene Aliases: ZNF548
UniProt ID: (Human) Q8NEK5
Entrez Gene ID: (Human) 147694
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support