Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide sequence: IKLEVYPTSL TKNYLEDSLL KILDRWIYGV SKNTFWSYFS QLQELCWEYY
Sequence homology: Human: 100%
The UGTs are of major importance in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds. UGT2B8 demonstrates reactivity with estriol. The UGTs are of major importance in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds. UGT2B8 demonstrates reactivity with estriol. See UGT2B4 (MIM 600067).[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HLUG4; UDP glucuronosyltransferase 2 family, member 15; UDP glucuronosyltransferase 2 family, member B8; UDP glucuronosyltransferase 2 family, polypeptide B15; UDP glycosyltransferase 2B15; UDP gycosyltransferase 2 family, member B15; UDP-glucuronosyltransferase 2B15; UDP-glucuronosyltransferase 2B8; UDP-glucuronosyltransferase UGT2B15; UDP-glucuronyltransferase, family 2, beta-15; UDPGT 2B15; UDPGT 2B8; UDPGTh-3; unnamed protein product; uridine diphosphate glucuronosyltransferase 2 family, member B8; uridine diphosphate glycosyltransferase 2 family, member B15
Gene Aliases: HLUG4; UDPGT 2B8; UDPGT2B15; UDPGTH3; UGT2B15; UGT2B8
UniProt ID: (Human) P54855
Entrez Gene ID: (Human) 7366
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support