Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide sequence: AQQPGRRGEG LSSPGGWLEA EKKGAPQKDS TGGVLEESNK IEPSLHSLQK
Sequence homology: Human: 100%
METTL21C is a protein-lysine methyltransferase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: methyltransferase like 21C; Methyltransferase-like protein 21C; Protein-lysine methyltransferase METTL21C
Gene Aliases: C13orf39; METTL21C
UniProt ID: (Human) Q5VZV1
Entrez Gene ID: (Human) 196541
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support