Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • WNT7A/WNT7B Antibodies

          Invitrogen

          WNT7A/WNT7B Polyclonal Antibody

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite WNT7A/WNT7B Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          WNT7A/WNT7B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          WNT7A/WNT7B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          WNT7A/WNT7B Antibody (PA5-47430) in IHC (P)

          Immunohistochemical analysis of WNT7A/WNT7B in immersion fixed paraffin-embedded sections of human endometrial cancer tissue. Samples were incubated in WNT7A/WNT7B polyclonal antibody (Product # PA5-47430) using a dilution of 15 µg/mL overnight at 4 °C. Tissue was stained with the Anti-Goat HRP-DAB Cell & Tissue Staining Kit (brown) and counterstained with hematoxylin (blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          WNT7A/WNT7B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          WNT7A/WNT7B Polyclonal Antibody

          Product Details

          PA5-47430

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          5-15 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Goat / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E. coli-derived recombinant human Wnt-7a Leu32-Glu72, Thr216-Arg292
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 5% trehalose

          Contains

          No Preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2610494

          Product Specific Information

          In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse (rm) Wnt-1, rmWnt-3a, rmWnt-4, rmWnt-5a, rmWnt-5b, rmWnt-9a, rmWnt-11, and rmWnt-16 is observed.

          Reconstitute at 0.2 mg/mL in sterile PBS.

          Target Information

          Wnt proteins constitute a family of secreted glycoproteins that activate signal transduction pathways to mediate tissue homeostasis, cell fate, cell proliferation, and self-renewal. The canonical signaling pathway is initiated upon Wnt binding to a Frizzled family receptor and an LRP5/LRP6 coreceptor, which leads to activation of the beta-catenin cascade. Non-canonical Wnt signals are activated by binding to a Frizzled family receptor and ROR2 coreceptor. Extensive research has shown that deregulation of these pathways or its components can lead to cancer. Human Wnt-1 is encoded as a 370 amino acid precursor that, upon secretion, is cleaved into a 343 amino acid (42 kDa) mature form. At the amino acid level, mature human Wnt-1 is approximately 99% identical to mature mouse Wnt-1. Depending on the tissue, Wnt-1 can activate the canonical and non-canonical signaling pathways. Studies have demonstrated its involvement in a variety of functions, including embryonic development and carcinogenesis. Wnt-1 signaling can also induce the phosphorylation and subsequent degradation of c-Myb.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Protein Wnt-7a; Protein Wnt-7b; proto-oncogene Wnt7a protein; unnamed protein product; wingless-type MMTV integration site family, member 7A; wingless-type MMTV integration site family, member 7B; WNT7a

          View more View less

          Gene Aliases: SANTOS; Wnt-7a; WNT7A; WNT7B

          View more View less

          UniProt ID: (Human) O00755, (Human) P56706

          View more View less

          Entrez Gene ID: (Human) 7476, (Human) 7477

          View more View less

          Function(s)
          receptor binding frizzled binding cytokine activity protein binding receptor agonist activity
          Process(es)
          embryonic axis specification cartilage condensation angiogenesis chondrocyte differentiation apoptotic process neurotransmitter secretion axonogenesis synapse assembly sex differentiation positive regulation of cell proliferation tissue development dorsal/ventral pattern formation positive regulation of endothelial cell migration positive regulation of gene expression skeletal muscle satellite cell activation skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration Wnt signaling pathway cerebellar granule cell differentiation cell proliferation in forebrain central nervous system vasculogenesis establishment of cell polarity cell differentiation neuron differentiation embryonic limb morphogenesis regulation of axon diameter response to estradiol somatic stem cell population maintenance embryonic forelimb morphogenesis embryonic hindlimb morphogenesis wound healing, spreading of epidermal cells non-canonical Wnt signaling pathway synaptic vesicle recycling regulation of cell proliferation embryonic digit morphogenesis negative regulation of apoptotic process response to estrogen cell fate commitment asymmetric protein localization involved in cell fate determination positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of JNK cascade somatic stem cell division animal organ development system development stem cell development regulation of epithelial cell proliferation negative regulation of neurogenesis regulation of axonogenesis synapse organization cartilage development positive regulation of protein metabolic process positive regulation of synapse assembly positive regulation of epithelial cell proliferation involved in wound healing uterus development oviduct development canonical Wnt signaling pathway Wnt signaling pathway, planar cell polarity pathway limb development establishment of blood-brain barrier dendritic spine morphogenesis uterus morphogenesis secondary palate development lens fiber cell development cellular response to transforming growth factor beta stimulus presynapse assembly postsynapse assembly regulation of postsynapse organization excitatory synapse assembly positive regulation of excitatory synapse assembly positive regulation of protein localization to presynapse regulation of presynapse assembly regulation of synaptic vesicle exocytosis positive regulation of excitatory postsynaptic potential in utero embryonic development metanephros morphogenesis establishment or maintenance of polarity of embryonic epithelium forebrain regionalization lung development neuron projection development oxygen homeostasis regulation of cell projection size chemoattraction of dopaminergic neuron axon homeostatic process positive regulation of osteoblast differentiation fibroblast proliferation embryonic organ development neuron projection morphogenesis response to glucocorticoid lung morphogenesis lung epithelium development lobar bronchus development trachea cartilage morphogenesis developmental growth involved in morphogenesis embryonic placenta morphogenesis chorio-allantoic fusion mammary gland epithelium development cellular response to retinoic acid renal inner medulla development renal outer medulla development outer medullary collecting duct development inner medullary collecting duct development stem cell proliferation metanephric collecting duct development metanephric epithelium development metanephric loop of Henle development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline