Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • IL10RB Antibodies

          Invitrogen

          IL10RB Polyclonal Antibody

          Advanced Verification
          View all (23) IL10RB antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite IL10RB Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Advanced Verification (1)
          IL10RB Antibody in Neutralization (Neu)
          Group 53 Created with Sketch.
          IL10RB Antibody in Neutralization (Neu)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IL10RB Antibody (PA5-47956) in Neu

          Neutralization of IL10RB in LPS-activated human peripheral blood mononuclear cells (PBMC). Samples were incubated in IL10RB polyclonal antibody (Product # PA5-47956). Recombinant Human IL‚10 inhibits IL‚1β secretion in LPS-activated human peripheral blood mononuclear cells (PBMC) in a dose-dependent manner (orange line), as measured by the Human IL‚1β/IL‚1F2 Quantikine ELISA Kit.IL‚1β secretion inhibited by Recombinant Human IL‚10 (0.25 ng/mL) is neutralized (green line) by increasing concentrations of Goat Anti-Human IL‚10 Rβ Antigen Affi... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IL10RB Antibody in Neutralization (Neu)
          IL10RB Antibody
          IL10RB Polyclonal Antibody

          Product Details

          PA5-47956

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1 µg/mL
          -

          Flow Cytometry (Flow)

          2.5 µg per million cells
          -

          Neutralization (Neu)

          2-6 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Goat / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Mouse myeloma cell line NS0-derived recombinant human IL-10 R beta Met20-Ser220
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 5% trehalose

          Contains

          No Preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2608054

          Product Specific Information

          In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) IL-20 R alpha, rhIL-10 R, and rhIL-20 R beta is observed.

          Reconstitute at 0.2 mg/mL in sterile PBS.

          Endoxin level is <0.20 EU per 1 µg of the antibody by the LAL method.

          Target Information

          Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CDw210b; CRF2-4; cytokine receptor class-II CRF2-4; Cytokine receptor class-II member 4; Cytokine receptor family 2 member 4; cytokine receptor family II, member 4; IL-10 receptor subunit beta; IL-10R subunit 2; IL-10R subunit beta; inte; interleukin 10 receptor, beta; Interleukin-10 receptor subunit 2; Interleukin-10 receptor subunit beta; unnamed protein product

          View more View less

          Gene Aliases: CDW210B; CRF2-4; CRFB4; D21S58; D21S66; IBD25; IL-10R2; IL-10RB; IL10RB

          View more View less

          UniProt ID: (Human) Q08334

          View more View less

          Entrez Gene ID: (Human) 3588

          View more View less

          Function(s)
          interleukin-10 receptor activity protein binding coreceptor activity signaling receptor activity
          Process(es)
          inflammatory response immune response signal transduction cytokine-mediated signaling pathway type III interferon-mediated signaling pathway positive regulation of JAK-STAT cascade defense response to virus cellular response to virus interleukin-10-mediated signaling pathway positive regulation of cellular respiration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline