Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
May have a role in a signaling pathway. Could act as a tumor suppressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Proline-glutamate repeat-containing protein; putative adapter and scaffold protein 1; SAM and SH3 domain-containing protein 1; unnamed protein product
Gene Aliases: 1100001C18Rik; 2500002E12Rik; A330076K04Rik; CAPOK; dJ323M4.1; DUH; DUH1; KIAA0790; mKIAA0790; PEPE1; SASH1; SH3D6A
UniProt ID: (Human) O94885, (Mouse) P59808
Entrez Gene ID: (Human) 23328, (Mouse) 70097
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support