Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Neurexin-1-beta (NRXN1-beta) is a gene that encodes the beta-isoform of the neurexin-1 protein, which is a type I membrane protein that functions as a cell adhesion molecule and receptor in the nervous system. Located on chromosome 2 in humans, the neurexin-1 gene undergoes complex alternative splicing, producing multiple isoforms, including the well-studied alpha-neurexin and beta-neurexin. Neurexins, including NRXN1-beta, are primarily involved in synapse formation and function by mediating interactions between presynaptic and postsynaptic neurons. They bind to neuroligins, another family of cell adhesion molecules, thus playing a critical role in the maintenance and regulation of synaptic specificity and neurotransmitter release. Dysregulation or mutations in NRXN1-beta have been implicated in various neurological and psychiatric disorders, including autism spectrum disorders, schizophrenia, and intellectual disabilities, emphasizing the importance of this gene in brain development and function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-latrotoxin receptor (calcium-dependent); neurexin 1; neurexin I; Neurexin I-alpha; Neurexin I-beta; Neurexin-1; Neurexin-1-alpha; Neurexin-1-beta; neuronal cell surface protein; non-processed neurexin I-alpha; unnamed protein product
Gene Aliases: 1700062G21Rik; 9330127H16Rik; A230068P09Rik; Hs.22998; KIAA0578; mKIAA0578; NRXN1; Nrxn1b; PTHSL2; SCZD17
UniProt ID: (Human) Q9ULB1, (Mouse) Q9CS84, (Rat) Q63372
Entrez Gene ID: (Human) 9378, (Mouse) 18189, (Rat) 60391
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support