Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The antibody detects endogenous levels of total NCEH1 protein.
Neutral cholesterol ester hydrolase 1 (NCEH) also known as arylacetamide deacetylase-like 1 (AADACL1) or KIAA1363 is an enzyme that in humans is encoded by the NCEH1 gene. NCEH is an enzyme located in the endoplasmic reticulum. NCEH hydrolyzes 2-acetyl monoalkylglycerol ether, as part of an enzymatic pathway regulating the levels of platelet activating factor and lysophospholipids that may be involved in cancer development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-acetyl MAGE hydrolase; Acetylalkylglycerol acetylhydrolase; alkylacetylglycerol acetylhydrolase; Arylacetamide deacetylase-like 1; Chlorpyrifos oxon-binding protein; CPO-BP; NCEH; Neutral cholesterol ester hydrolase 1; unnamed protein product
Gene Aliases: AADACL1; B230106I24Rik; CPO-BP; KIAA1363; NCEH; NCEH1
UniProt ID: (Human) Q6PIU2, (Mouse) Q8BLF1
Entrez Gene ID: (Human) 57552, (Mouse) 320024
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support