Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
NTE Polyclonal Antibody detects endogenous levels of NTE protein.
This gene encodes a phospholipase that deacetylates intracellular phosphatidylcholine to produce glycerophosphocholine. It is thought to function in neurite outgrowth and process elongation during neuronal differentiation. The protein is anchored to the cytoplasmic face of the endoplasmic reticulum in both neurons and non-neuronal cells. Mutations in this gene result in autosomal recessive spastic paraplegia, and the protein is the target for neurodegeneration induced by organophosphorus compounds and chemical warfare agents. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD73; Neuropathy target esterase; patatin like phospholipase domain containing 6; patatin-like phospholipase domain containing 6; Patatin-like phospholipase domain-containing protein 6; Swiss cheese; Swiss-cheese; unnamed protein product
Gene Aliases: AI661849; BNHS; iPLA2delta; LNMS; MSws; NTE; NTEMND; OMCS; PNPLA6; RGD1564611; SPG39; sws
UniProt ID: (Human) Q8IY17, (Mouse) Q3TRM4
Entrez Gene ID: (Human) 10908, (Mouse) 50767, (Rat) 360753
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support