Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: RKAGLAHLYG IAGSVNVTGD EVKKLDVLSN SLVINMVQSS YSTCVLVSEE NKDAIITAKE KRGKYVVCFD PLDG
This product detects both FBP1 and FBP2, which are 80% homologous. The antigen sequence is a 100% match to FBP2 and the product has been validated against FBP1.
Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.
FBP1 and FBP2 are gluconeogenesis regulatory enzymes which catalyze the hydrolosis of fructose 1,6-bisphosphate to fructose 6-phosphate and inorganic phosphate. Fructose-1,6-diphosphatase deficiency is associated with hypoglycemia and metabolic acidosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D-fructose-1,6-bisphosphate 1-phosphohydrolase 1; D-fructose-1,6-bisphosphate 1-phosphohydrolase 2; FBP2; FBPase 1; FBPase 2; fructose 1,6-bisphosphatase (EC 3.1.3.11); fructose-1; Fructose-1,6-bisphosphatase 1; fructose-1,6-bisphosphatase 2; Fructose-1,6-bisphosphatase isozyme 2; GIG17; growth-inhibiting protein 17; hexosediphosphatase; Liver FBPase; Muscle FBPase; muscle fructose-bisphosphatase; unnamed protein product
Gene Aliases: CORLK; FBP; FBP1; FBP2
UniProt ID: (Human) P09467, (Human) O00757
Entrez Gene ID: (Human) 2203, (Human) 8789
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support