Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: GNVRVKFGQN DADSSPGRDY EEDEIKNHAS DRRQKGQHHE RTGQDGHLYG SSPVEREEGA LRRATTTARS LLDNKVVKNS RADGGRLAGE GQLQNIPIDF TNFPAHVDLP KAGSGLEEND LVQ
Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 62%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: protein phosphatase 1, regulatory subunit 152; Transmembrane protein 132C; unnamed protein product
Gene Aliases: PPP1R152; TMEM132C
UniProt ID: (Human) Q8N3T6
Entrez Gene ID: (Human) 92293
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support