Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: NDLTFTEEVD MKEMEKALQE RDYIFGNYIE RLWAYLTIEQ LLEKRKNAHG EEKENLTARA LDLSLKYHFV TPLTSMVVTK PEDNEDERAI ADKPGEDAEA TPVSPAMSYL TSYQPPQ
Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 77%.
This gene encodes the heavy chain subunit of the pre-alpha-trypsin inhibitor complex. This complex may stabilize the extracellular matrix through its ability to bind hyaluronic acid. Polymorphisms of this gene may be associated with increased risk for schizophrenia and major depressive disorder. This gene is present in an inter-alpha-trypsin inhibitor family gene cluster on chromosome 3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: inter-alpha (globulin) inhibitor H3; inter-alpha (globulin) inhibitor, H3 polypeptide; inter-alpha-inhibitor heavy chain 3; Inter-alpha-trypsin inhibitor heavy chain H3; ITI heavy chain H3; pre-alpha (globulin) inhibitor, H3 polypeptide; Serum-derived hyaluronan-associated protein; SHAP; unnamed protein product
Gene Aliases: H3P; ITI-HC3; ITIH3; SHAP
UniProt ID: (Human) Q06033
Entrez Gene ID: (Human) 3699
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support