Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: NTQGCTTEGD LLFAQKCKEL QGFIPPLTDL LNGLKMGRFE RGLSSFQQSV AMDRIQRIVG VLQKPQMGER YLGTLLQVEG MLKTWFPQ
Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 95%.
Transcriptional repressor which forms a negative regulatory component of the circadian clock and acts independently of the circadian transcriptional repressors: CRY1, CRY2 and BHLHE41. In a histone deacetylase-dependent manner represses the transcriptional activator activity of the CLOCK-ARNTL/BMAL1 heterodimer. Abrogates the interaction of ARNTL/BMAL1 with the transcriptional coactivator CREBBP and can repress the histone acetyl-transferase activity of the CLOCK-ARNTL/BMAL1 heterodimer, reducing histone acetylation of its target genes. Rhythmically binds the E-box elements (5'-CACGTG-3') on circadian gene promoters and its occupancy shows circadian oscillation antiphasic to ARNTL/BMAL1. Interacts with the glucocorticoid receptor (NR3C1) and contributes to the repressive function in the glucocorticoid response. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ChIP-derived repressor of network oscillator; Chrono; circadian associated repressor of transcription; Circadian-associated transcriptional repressor; Computationally highlighted repressor of the network oscillator; hypothetical gene supported by NM_017187; unnamed protein product
Gene Aliases: C1orf51; CHRONO; CIART; GM129; RGD1566380
UniProt ID: (Human) Q8N365, (Mouse) Q3TQ03
Entrez Gene ID: (Human) 148523, (Mouse) 229599, (Rat) 365871
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support