Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SELNHVKVTS EALGHAGRHL QQIILDQCYN FVCVNVTTSD FQETQKLLSM LEESSLCILY PVVVVSVHFL DFKLVPPSQK MENLMQ
Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 58%.
Required for protein O-linked mannosylation. Probably acts as a nucleotidyltransferase involved in synthesis of a nucleotide sugar. Required for dystroglycan O-mannosylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-C-methyl-D-erythritol 4-phosphate cytidylyltransferase-like protein; 4-diphosphocytidyl-2C-methyl-D-erythritol synthase homolog; D-ribitol-5-phosphate cytidylyltransferase; hISPD; isoprenoid synthase domain containing; Isoprenoid synthase domain-containing protein; notch1-induced protein; testicular tissue protein Li 97; unnamed protein product
Gene Aliases: CRPPA; hISPD; ISPD; LGMDR20; MDDGA7; MDDGC7; Nip
UniProt ID: (Human) A4D126
Entrez Gene ID: (Human) 729920
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support