Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: RQVLYPRNST GAYCGMGENK DKPYLLYFNI FSCILSSNII SVAENGLQCP TPQVCVSSCP EDPWTVGKNE FSQTVGEVFY TKNRNFCLPG V
Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 71%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Choline transporter-like protein 4; hTPPT1; SLC44A4 (hTPPT); Solute carrier family 44 member 4; testicular tissue protein Li 48; Thiamine pyrophosphate transporter 1; unnamed protein product
Gene Aliases: C6orf29; CTL4; DFNA72; hTPPT1; NG22; SLC44A4; TPPT; TPPT1; UNQ441/PRO874
UniProt ID: (Human) Q53GD3
Entrez Gene ID: (Human) 80736
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support