Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: QLHCRDVVQM VAQLVSEDVD KDVLLPHPLR STESTNAFQA FLARSAPFWH NATFEAS
Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 47%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: testis expressed gene 22; Testis-expressed protein 22; testis-expressed sequence 22 protein
Gene Aliases: TEX22
UniProt ID: (Human) C9J3V5
Entrez Gene ID: (Human) 647310
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support