Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This target displays homology in the following species: Cow: 92%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 87%; Rabbit: 93%; Rat: 86%
ZFHX2 gene ontology annotations related to this gene include adult behavior; regulation of sensory perception of pain; regulation of transcription, DNA-templated.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ZFH-2; Zinc finger homeobox protein 2; Zinc finger homeodomain protein 2; zinc finger protein 409
Gene Aliases: KIAA1056; KIAA1762; MARSIS; ZFH-5; ZFH5; ZFHX2; ZNF409
UniProt ID: (Human) Q9C0A1
Entrez Gene ID: (Human) 85446
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support